2,6-Difluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3-Difluoro-2-methylbenzene
B25076-06
B25076-14
CAAAAB25076-03EA
56.94
CAD
CAAAAB25076-03
CAAAAB25076-06
CAAAAB25076-14
2,6-Difluorotoluene 98%
2,6-Difluorotoluene
Purity: 98%
CAS No.: 443-84-5
Molecular Formula: C7H6F2
Formula Weight: 128.12
Flash Point: 10°(50°F)
Refractive Index: 1.4528
Odor: Aromatic
Boiling Point: 112-115°
Beilstein Reference: 1932656
UN No.: 2388
MDL No.: MFCD00043898
Formula:
C₇H₆F₂ Boiling Pt: 112…115 °C Density: 1.130 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043898 CAS Number: 443-84-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...