2-Bromo-5-fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25071-06
B25071-14
CAAAAB25071-06EA
114.39
CAD
CAAAAB25071-06
CAAAAB25071-14
2-Bromo-5-fluorobenzaldehyde 98%
2-Bromo-5-fluorobenzaldehyde
Purity: 98%
CAS No.: 94569-84-3
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Flash Point: 87°(190°F)
Melting Point: 51-56°
MDL No.: MFCD00142872
Formula:
C₇H₄BrFO Melting Pt: 51…56 °C Flash Pt: 87 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142872 CAS Number: 94569-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...