1-Propyl-4-ethynylbenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne
B25065-06
B25065-14
CAAAAB25065-06EA
222.78
CAD
CAAAAB25065-06
CAAAAB25065-14
CAAAAB25065-22
1-Propyl-4-ethynylbenzene 97%
1-Propyl-4-ethynylbenzene
Purity: 97%
CAS No.: 62452-73-7
Molecular Formula: C11H12
Formula Weight: 144.22
Flash Point: 67°(153°F)
Refractive Index: 1.5340
Boiling Point: 205°
MDL No.: MFCD00173886
Formula:
C₁₁H₁₂ MW: 144.22 g/mol Boiling Pt: 205 °C Density: 0.91 Flash Pt: 67 °C(153 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00173886 CAS Number: 62452-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...