α-Bromo-3-(trifluoromethoxy)toluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25061-03
B25061-06
CAAAAB25061-03EA
43.29
CAD
CAAAAB25061-03
CAAAAB25061-06
α-Bromo-3-(trifluoromethoxy)toluene 98%
α-Bromo-e-(trifluoromethoxy)toluene
Purity: 98%
CAS No.: 159689-88-0
Molecular Formula: C8H6BrF3O
Formula Weight: 255.04
Flash Point: 88°(190°F)
Refractive Index: 1.4780
Boiling Point: 178-180°
Beilstein Reference: 8052203
UN No.: 3265
MDL No.: MFCD00061271
Formula:
C₈H₆BrF₃O Boiling Pt: 178…180 °C Density: 1.572 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061271 CAS Number: 159689-88-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...