3,4-Dimethoxybenzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25033-06
B25033-22
CAAAAB25033-06EA
132.56
CAD
CAAAAB25033-06
CAAAAB25033-22
3,4-Dimethoxybenzylamine 97%
3,4-Dimethoxybenzylamine
Purity: 97%
CAS No.: 5763-61-1
Molecular Formula: C9H13NO2
Formula Weight: 167.21
Flash Point:
Formula:
C₉H₁₃NO₂ Boiling Pt: 281…284 °C Density: 1.109 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008116 CAS Number: 5763-61-1 EINECS: 227-287-7 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...