Methyl-3-nitrosalicylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-hydroxy-3-nitrobenzoate

B25029-06 B25029-14
CAAAAB25029-03EA 77.78 CAD
CAAAAB25029-03 CAAAAB25029-06 CAAAAB25029-14
Methyl-3-nitrosalicylate 98%
Methyl 3-nitrosalicylate

Purity: 98% CAS No.: 22621-41-6 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 129-132° MDL No.: MFCD00272263

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00272263
CAS Number: 22621-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR