6-Hydroxyflavone 98%
Supplier: Thermo Scientific Chemicals
B25016-03
B25016-06
CAAAAB25016-03EA
66.52
CAD
CAAAAB25016-03
CAAAAB25016-06
CAAAAB25016-14
6-Hydroxyflavone 98%
6-Hydroxyflavone
Purity: 98%
CAS No.: 6665-83-4
Molecular Formula: C15H10O3
Formula Weight: 238.25
Melting Point: 234-236°
Beilstein Reference: 15827
Formula:
C₁₅H₁₀O₃ Boiling Pt: 395 °C (1013 hPa) Melting Pt: 234…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017329 CAS Number: 6665-83-4 EINECS: 229-704-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...