6-Methylpyridin-3-ol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Methylpyridin-5-ol, 3-Hydroxy-6-methylpyridine, 5-Hydroxy-2-methylpyridine

B25014-06 B25014-14 B25014-22
CAAAAB25014-06EA 41.08 CAD
CAAAAB25014-06 CAAAAB25014-14 CAAAAB25014-22
6-Methylpyridin-3-ol 99%
6-Methylpyridin-3-ol

Purity: 99% CAS No.: 1121-78-4 Molecular Formula: C6H7NO Formula Weight: 109.13 Odor: Odorless Melting Point: 167-171° Beilstein Reference: 107077 MDL No.: MFCD00006339

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00006339
CAS Number: 1121-78-4
EINECS: 214-337-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR