Pinacol, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Dimethylbutane-2,3-diol, Tetramethylethylene glycol, 2,3-Dimethyl-2,3-butanediol
B25006-14
B25006-22
B25006-36
CAAAAB25006-14EA
80.58
CAD
CAAAAB25006-14
CAAAAB25006-22
CAAAAB25006-36
Pinacol, anhydrous 99%
Pinacol
Purity: 99%
CAS No.: 76-09-5
Molecular Formula: C6H14O2
Formula Weight: 118.18
Flash Point: 77°(170°F)
Melting Point: 42-45°
Boiling Point: 171-172°
Beilstein Reference: 1340501
Merck Reference: 14,7439
MDL No.: MFCD00004462
Formula:
C₆H₁₄O₂ Boiling Pt: 171…172 °C Melting Pt: 42…45 °C Density: 0.967 g/cm³ (20 °C) Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004462 CAS Number: 76-09-5 EINECS: 200-933-5 Merck Index: 14,7439 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...