2-Bromo-1,3,5-trichlorobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

B25000-06 B25000-14
CAAAAB25000-06EA 164.32 CAD
CAAAAB25000-06 CAAAAB25000-14
2-Bromo-1,3,5-trichlorobenzene 97%
2-Bromo-1,3,5-trichlorobenzene

Purity: 97% CAS No.: 19393-96-5 Molecular Formula: C6H2BrCl3 Formula Weight: 260.34 MDL No.: MFCD00060653

Formula: C₆H₂BrCl₃
Storage Temperature: Ambient
MDL Number: MFCD00060653
CAS Number: 19393-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR