2-Fluorothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

B24995-03 B24995-06 B24995-14
CAAAAB24995-03EA 42.66 CAD
CAAAAB24995-03 CAAAAB24995-06 CAAAAB24995-14
2-Fluorothiophenol 97%
2-Fluorothiophenol

Purity: 97% CAS No.: 2557-78-0 Molecular Formula: C6H5FS Formula Weight: 128.17 Flash Point: 56°(133°F) Refractive Index: 1.5550 Odor: Stench Boiling Point: 164° Beilstein Reference: 2039768 UN No.: 3071 MDL No.: MFCD00041419

Formula: C₆H₅FS
Boiling Pt: 164 °C
Density: 1.203
Flash Pt: 56 °C (133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041419
CAS Number: 2557-78-0
UN: 1228
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR