4-Fluorosalicylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24992-03
B24992-06
B24992-14
CAAAAB24992-03EA
90.69
CAD
CAAAAB24992-03
CAAAAB24992-06
CAAAAB24992-14
4-Fluorosalicylic acid 98%
4-Fluorosalicylic acid
Purity: 98%
CAS No.: 345-29-9
Molecular Formula: C7H5FO3
Formula Weight: 156.11
Melting Point: 186-190°
Beilstein Reference: 2209124
Formula:
C₇H₅FO₃ Melting Pt: 186…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153168 CAS Number: 345-29-9 EINECS: 206-459-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...