2-Bromo-5-fluorobenzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24989-06
B24989-14
CAAAAB24989-06EA
53.09
CAD
CAAAAB24989-06
CAAAAB24989-14
2-Bromo-5-fluorobenzonitrile 98+%
2-Bromo-5-fluorobenzonitrile
Purity: 98+%
CAS No.: 57381-39-2
Molecular Formula: C7H3BrFN
Formula Weight: 200.01
Melting Point: 88-92°
Beilstein Reference: 8678330
UN No.: 3439
MDL No.: MFCD00142875
Formula:
C₇H₃BrFN Boiling Pt: 255 °C (1013 hPa) Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142875 CAS Number: 57381-39-2 EINECS: 260-711-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...