1,1-Dimethoxyacetone 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
B24988-22
B24988-36
CAAAAB24988-22EA
86.27
CAD
CAAAAB24988-22
CAAAAB24988-36
1,1-Dimethoxyacetone 97+%
1,1-Dimethoxyacetone
Purity: 97+%
CAS No.: 6342-56-9
Molecular Formula: CH3COCH(OCH3)2
Formula Weight: 118.13
Flash Point: 34°(93°F)
Refractive Index: 1.3980
Melting Point: -57°
Boiling Point: 133-135°
Beilstein Reference: 1560557
UN No.: 1993
MDL No.: MFCD00008758
Formula:
C₅H₁₀O₃ Boiling Pt: 133…135 °C Melting Pt: –57 °C Density: 0.998 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008758 CAS Number: 6342-56-9 EINECS: 228-735-4 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...