5-Amino-4,6-dichloropyrimidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,6-Dichloropyrimidin-5-ylamine

B24985-06 B24985-14
CAAAAB24985-03EA 83.04 CAD
CAAAAB24985-03 CAAAAB24985-06 CAAAAB24985-14
5-Amino-4,6-dichloropyrimidine 98+%
5-Amino-4,6-dichloropyrimidine

Purity: 98+% CAS No.: 5413-85-4 Molecular Formula: C4H3Cl2N3 Formula Weight: 164.00 Odor: Irritating Melting Point: 143-146° Beilstein Reference: 126885 MDL No.: MFCD00006108

Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
Boiling Pt: 262
Melting Pt: 147
Storage Temperature: Ambient
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR