Octyl butyrate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Butanoic acid octyl ester, Butyric acid octyl ester

B24978-18 B24978-30
CAAAAB24978-0BEA 533.49 CAD
CAAAAB24978-0B CAAAAB24978-18 CAAAAB24978-30
Octyl butyrate 97%
Octyl butyrate

Purity: 97% CAS No.: 110-39-4 Molecular Formula: CH3CH2CH2CO2(CH2)7CH3 Formula Weight: 172.27 MDL No.: MFCD00048939

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: –56 °C
Density: 0,87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00048939
CAS Number: 110-39-4
EINECS: 203-762-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR