2,6-Dimethyl-4-fluoroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluoro-2,6-xylidine, 2-Amino-5-fluoro-m-xylene

B24965-03 B24965-06 B24965-14
CAAAAB24965-03EA 210.14 CAD
CAAAAB24965-03 CAAAAB24965-06 CAAAAB24965-14
2,6-Dimethyl-4-fluoroaniline 98%
2,6-Dimethyl-4-fluoroaniline

Purity: 98% CAS No.: 392-70-1 Molecular Formula: C8H10FN Formula Weight: 139.17 Refractive Index: 1.5345

Formula: C₈H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD03792669
CAS Number: 392-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR