5-Nitroisatin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24962-06
B24962-14
CAAAAB24962-06EA
63.99
CAD
CAAAAB24962-06
CAAAAB24962-14
CAAAAB24962-22
5-Nitroisatin 98%
5-Nitroisatin
Purity: 98%
CAS No.: 611-09-6
Molecular Formula: C8H4N2O4
Formula Weight: 192.13
Melting Point: ca 252° dec.
Beilstein Reference: 180223
Formula:
C₈H₄N₂O₄ Melting Pt: ∼ 252 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005720 CAS Number: 611-09-6 EINECS: 210-252-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...