Sodium-2-ethylhexanoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24960-0B
B24960-0I
B24960-30
CAAAAB24960-0BEA
169.06
CAD
CAAAAB24960-0B
CAAAAB24960-0I
CAAAAB24960-30
Sodium-2-ethylhexanoate 97%
Sodium 2-ethylhexanoate
Purity: 97%
CAS No.: 19766-89-3
Molecular Formula: C8H15NaO2
Formula Weight: 166.20
Odor: Odorless
Melting Point:
Formula:
C₈H₁₅NaO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014007 CAS Number: 19766-89-3 EINECS: 243-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...