α,α'-Dibromo-m-xylene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

B24959-09 B24959-18 B24959-30
CAAAAB24959-09EA 118.82 CAD
CAAAAB24959-09 CAAAAB24959-18 CAAAAB24959-30
α,α'-Dibromo-m-xylene 97%
α,α'-Dibromo-m-xylene

Purity: 97% CAS No.: 626-15-3 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 74-76° Beilstein Reference: 971085 UN No.: 3261 MDL No.: MFCD00000178

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 135 - 140 °C (20 -rr)
Melting Pt: 74…76 °C
Density: 1.959 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000178
CAS Number: 626-15-3
EINECS: 210-931-6
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR