2-Methylpyridin-3-ol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24944-03
B24944-06
B24944-14
CAAAAB24944-03EA
50.56
CAD
CAAAAB24944-03
CAAAAB24944-06
CAAAAB24944-14
2-Methylpyridin-3-ol 99%
2-Methylpyridin-3-ol
Purity: 99%
CAS No.: 1121-25-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Odor: Odorless
Melting Point: 166-169°
Beilstein Reference: 107937
MDL No.: MFCD00082538
Formula:
C₆H₇NO Melting Pt: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082538 CAS Number: 1121-25-1 EINECS: 214-327-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...