3,4-Dimethylbenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Hydroxymethyl)-o-xylene, (3,4-Dimethylphenyl)methanol

B24926-03 B24926-06 B24926-14
CAAAAB24926-03EA 61.94 CAD
CAAAAB24926-03 CAAAAB24926-06 CAAAAB24926-14
3,4-Dimethylbenzyl alcohol 97%
3,4-Dimethylbenzyl alcohol

Purity: 97% CAS No.: 6966-10-5 Molecular Formula: C9H12O Formula Weight: 136.19 Melting Point: 62-65° Boiling Point: 218-221° MDL No.: MFCD00004647

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 218
Melting Pt: 63
Storage Temperature: Ambient
MDL Number: MFCD00004647
CAS Number: 6966-10-5
EINECS: 230-175-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR