1-Benzyloxy-4-bromobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

B24923-03 B24923-06 B24923-14
CAAAAB24923-03EA 59.57 CAD
CAAAAB24923-03 CAAAAB24923-06 CAAAAB24923-14
1-Benzyloxy-4-bromobenzene 97%
1-Benzyloxy-4-bromobenzene

Purity: 97% CAS No.: 6793-92-6 Molecular Formula: C13H11BrO Formula Weight: 263.14 Melting Point: 61-63° Boiling Point: 166-167°/3mm MDL No.: MFCD00028016

Formula: C₁₃H₁₁BrO
Boiling Pt: 166…167 °C/3 mm
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00028016
CAS Number: 6793-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR