2-Fluorobenzhydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

B24921-09 B24921-18 B24921-30
CAAAB24921-09EA 79.63 CAD
CAAAB24921-09 CAAAB24921-18 CAAAB24921-30
2-Fluorobenzhydrazide 98%
2-Fluorobenzhydrazide

Purity: 98% CAS No.: 446-24-2 Molecular Formula: C7H7FN2O Formula Weight: 154.14 Melting Point: 72-74° MDL No.: MFCD00025112

Formula: C₇H₇FN₂O
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00025112
CAS Number: 446-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR