3-Nitro-o-xylene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24917-22
B24917-36
CAAAAB24917-22EA
61.62
CAD
CAAAAB24917-22
CAAAAB24917-36
3-Nitro-o-xylene 99%
3-Nitro-o-xylene
Purity: 99%
CAS No.: 83-41-0
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 107°(224°F)
Refractive Index: 1.5440
Melting Point: 13-15°
Boiling Point: 244-246°
Beilstein Reference: 2045105
UN No.: 1665
MDL No.: MFCD00007162
Formula:
C₈H₉NO₂ Boiling Pt: 244…246 °C Melting Pt: 13…15 °C Density: 1.129 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007162 CAS Number: 83-41-0 EINECS: 201-474-3 UN: 1665 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...