(S)-(+)-2-Amino-1-propanol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol

B24916-06 B24916-14
CAAAAB24916-06EA 189.6 CAD
CAAAAB24916-06 CAAAAB24916-14
(S)-(+)-2-Amino-1-propanol 98%
(S)-(+)-2-Amino-1-propanol

Purity: 98% CAS No.: 2749-11-3 Molecular Formula: C3H9NO Formula Weight: 75.11 Flash Point: 83°(181°F) Refractive Index: 1.4498 Boiling Point: 174-176° Beilstein Reference: 1718865 Optical Rotation: +22° (c=2 in ethanol) UN No.: 2735 MDL No.: MFCD00064412

Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 174…176 °C
Density: 0.965
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR