4-Bromo-2-chlorobenzenesulfonyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Bromo-2-chlorobenzene-1-sulphonyl chloride, 4-Bromo-2-chlorobenzene-1-sulfonyl chloride
B24913-06
B24913-14
B24913-22
CAAAAB24913-06EA
162.74
CAD
CAAAAB24913-06
CAAAAB24913-14
CAAAAB24913-22
4-Bromo-2-chlorobenzenesulfonyl chloride 96%
4-Bromo-2-chlorobenzenesulphonyl chloride
Purity: 96%
CAS No.: 351003-52-6
Molecular Formula: C6H3BrCl2O2S
Formula Weight: 289.96
Melting Point: 76-81°
UN No.: 3261
MDL No.: MFCD03094630
Formula:
C₆H₃BrCl₂O₂S MW: 289.96 g/mol Melting Pt: 76…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094630 CAS Number: 351003-52-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...