2-Phenylbenzoxazole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenyl-1,3-benzoxazole, 2-Phenylbenzo[d]oxazole

B24906-03 B24906-06
CAAAAB24906-03EA 81.53 CAD
CAAAAB24906-03 CAAAAB24906-06 CAAAAB24906-14
2-Phenylbenzoxazole 99%
2-Phenylbenzoxazole

Purity: 99% CAS No.: 833-50-1 Molecular Formula: C13H9NO Formula Weight: 195.22 Melting Point: 102-104° MDL No.: MFCD00012183

Formula: C₁₃H₉NO
MW: 195.22 g/mol
Boiling Pt: 314…317 °C (1013 hPa)
Melting Pt: 102…104 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012183
CAS Number: 833-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR