2-Bromo-5-fluoroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

B24901-06 B24901-14
CAAAAB24901-06EA 89.74 CAD
CAAAAB24901-06 CAAAAB24901-14
2-Bromo-5-fluoroaniline 98%
2-Bromo-5-fluoroaniline

Purity: 98% CAS No.: 1003-99-2 Molecular Formula: C6H5BrFN Formula Weight: 190.01 Flash Point: 110°(230°F) Melting Point: 42-46° Boiling Point: 154°/23mm UN No.: 2811 MDL No.: MFCD00070750

Formula: C₆H₅BrFN
Boiling Pt: 154 °C/23 mm
Melting Pt: 42…46 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070750
CAS Number: 1003-99-2
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR