1-Chlorooctadecane 94%
Supplier: Thermo Scientific Chemicals
B24900-14
B24900-22
B24900-36
CAAAAB24900-14EA
60.36
CAD
CAAAAB24900-14
CAAAAB24900-22
CAAAAB24900-36
1-Chlorooctadecane 94%
1-Chlorooctadecane
Purity: 94%
CAS No.: 3386-33-2
Molecular Formula: C18H37Cl
Formula Weight: 288.95
Flash Point:
Formula:
C₁₈H₃₇Cl Boiling Pt: 157…158 °C/1.5 mm Melting Pt: 20…23 °C Density: 0.849 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000960 CAS Number: 3386-33-2 EINECS: 222-207-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...