Gallic acid, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Warning

B24887-0B B24887-30
CAAAAB24887-0BEA 472.42 CAD
CAAAAB24887-0B CAAAAB24887-30
Gallic acid, anhydrous 98%
Gallic acid

Organic acid used as a standard for determining phenols in the Folin-Ciocalteau assay


Purity: 99% CAS No.: 149-91-7 Molecular Formula: C7H6O5 Formula Weight: 170.12 Melting Point: ca 251° dec. Beilstein Reference: 2050274 Merck Reference: 14,4345 MDL No.: MFCD00002510

Formula: C₇H₆O₅
Boiling Pt: 501 °C (1013 hPa)
Melting Pt: ca 251 °C (dec.)
Density: 1.694
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 14,4345

Order Now

Specification Test Results

Appearance (Color) White to cream or pale yellow
Form Powder
Assay (aqueous acid-base titration) ≥97.5 to ≤102.5%
Assay (Silylated GC) ≥97.5 to ≤102.5%
Identification (FTIR) Conforms

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR