5-Bromo-1,2,3-trimethoxybenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24872-06
B24872-14
B24872-22
CAAAB24872-06EA
67.31
CAD
CAAAB24872-06
CAAAB24872-14
CAAAB24872-22
5-Bromo-1,2,3-trimethoxybenzene 97%
5-Bromo-1,2,3-trimethoxybenzene
Purity: 97%
CAS No.: 2675-79-8
Molecular Formula: C9H11BrO3
Formula Weight: 247.10
Melting Point: 77-80°
MDL No.: MFCD00017169
Formula:
C₉H₁₁BrO₃ Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017169 CAS Number: 2675-79-8 EINECS: 220-223-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...