Dithiooxamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethanebis(thioamide), Rubeanic acid

B24866-06 B24866-14
CAAAAB24866-06EA 154.84 CAD
CAAAAB24866-06 CAAAAB24866-14
Dithiooxamide 98%
Dithiooxamide

Purity: 98% CAS No.: 79-40-3 Molecular Formula: C2H4N2S2 Formula Weight: 120.2 Melting Point: 245° dec. MDL No.: MFCD00004941

Formula: C₂H₄N₂S₂
Melting Pt: 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004941
CAS Number: 79-40-3
EINECS: 201-203-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR