5-Bromo-2-chloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

B24862-06 B24862-14
CAAAAB24862-03EA 41.7 CAD
CAAAAB24862-03 CAAAAB24862-06 CAAAAB24862-14
5-Bromo-2-chloropyridine 98%
5-Bromo-2-chloropyridine

Purity: 98% CAS No.: 53939-30-3 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Melting Point: 68-72° Beilstein Reference: 108887

Formula: C₅H₃BrClN
MW: 192.44
Boiling Pt: 205
Melting Pt: 71
Storage Temperature: Ambient
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR