1-Chloro-3,5-dimethoxybenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chloro-1,3-dimethoxybenzene
B24834-06
B24834-14
B24834-22
CAAAAB24834-06EA
67.62
CAD
CAAAAB24834-06
CAAAAB24834-14
CAAAAB24834-22
1-Chloro-3,5-dimethoxybenzene 98%
1-Chloro-3,5-dimethoxybenzene
Purity: 98%
CAS No.: 7051-16-3
Molecular Formula: C8H9ClO2
Formula Weight: 172.61
Flash Point: 112°(233°F)
Melting Point: 32-36°
Boiling Point: 234°
Beilstein Reference: 2327304
MDL No.: MFCD00008382
Formula:
C₈H₉ClO₂ MW: 172.61 g/mol Boiling Pt: 234 Melting Pt: 38 Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008382 CAS Number: 7051-16-3 EINECS: 230-330-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...