(2-Bromoethyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24833-06
B24833-14
CAAAAB24833-06EA
97.96
CAD
CAAAAB24833-06
CAAAAB24833-14
CAAAAB24833-22
(2-Bromoethyl)triphenylphosphonium bromide 98%
(2-Bromoethyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 7301-93-1
Molecular Formula: BrCH2CH2P(C6H5)3Br
Formula Weight: 450.15
Melting Point: 186-189°
MDL No.: MFCD04039352
Formula:
C₂₀H₁₉Br₂P Melting Pt: 186…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039352 CAS Number: 7301-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...