1-Methoxynaphthalene 98+%

Supplier: Thermo Scientific Chemicals

B24825-14 B24825-22 B24825-36
CAAAAB24825-14EA 90.22 CAD
CAAAAB24825-14 CAAAAB24825-22 CAAAAB24825-36
1-Methoxynaphthalene 98+%
1-Methoxynaphthalene

Purity: 98+% CAS No.: 2216-69-5 Molecular Formula: C11H10O Formula Weight: 158.20 Flash Point: 110°(230°F) Refractive Index: 1.6210 Melting Point: 5° Boiling Point: 135-137°/12mm Beilstein Reference: 7748884 MDL No.: MFCD00003924

Formula: C₁₁H₁₀O
Boiling Pt: 135…137 °C/12 mm
Melting Pt: 5 °C
Density: 1.094
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003924
CAS Number: 2216-69-5
EINECS: 218-696-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR