Acrolein diethyl acetal 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24821-09
B24821-18
CAAAAB24821-09EA
92.27
CAD
CAAAAB24821-09
CAAAAB24821-18
Acrolein diethyl acetal 96%
Acrolein diethyl acetal
Purity: 96%
CAS No.: 3054-95-3
Molecular Formula: CH2=CHCH(OCH2CH3)2
Formula Weight: 130.19
Flash Point: 13°(55°F)
Refractive Index: 1.4010
Boiling Point: 124-125°
Beilstein Reference: 1701567
UN No.: 2374
MDL No.: MFCD00009239
Formula:
C₇H₁₄O₂ Boiling Pt: 124…125 °C Melting Pt: –77 °C Density: 0.852 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009239 CAS Number: 3054-95-3 EINECS: 221-276-0 UN: 2374 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...