2-Hydroxyethyl macrylate 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 200-300 ppm
B24815-0B
B24815-30
CAAAAB24815-0BEA
254.38
CAD
CAAAAB24815-0B
CAAAAB24815-30
2-Hydroxyethyl macrylate 97% stabilized
2-Hydroxyethyl macrylate
As a reactive monomer
Purity: 97%
CAS No.: 818-61-1
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 101°(214°F)
Refractive Index: 1.4500
Odor: Pungent
Melting Point:
Formula:
C₅H₈O₃ Boiling Pt: 192 °C Melting Pt: < –60 °C Density: 1.106 Flash Pt: 101 °C(214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002865 CAS Number: 818-61-1 EINECS: 212-454-9 UN: 2927 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...