1,3-Dimethoxy-5-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Fluoro-3,5-dimethoxybenzene

B24810-06 B24810-14 B24810-22
CAAAAB24810-06EA 105.86 CAD
CAAAAB24810-06 CAAAAB24810-14 CAAAAB24810-22
1,3-Dimethoxy-5-fluorobenzene 98%
1,3-Dimethoxy-5-fluorobenzene

Purity: 98% CAS No.: 52189-63-6 Molecular Formula: C8H9FO2 Formula Weight: 156.16 Flash Point: 91°(196°F) Refractive Index: 1.5025 Boiling Point: 86-88°/7mm MDL No.: MFCD00012445

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Boiling Pt: 86…88 °C (7 mmHg)
Density: 1.170
Flash Pt: 91 °C (196 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012445
CAS Number: 52189-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR