4-Bromo-2-chlorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

B24803-09 B24803-18 B24803-30
CAAAAB24803-09EA 81.69 CAD
CAAAAB24803-09 CAAAAB24803-18 CAAAAB24803-30
4-Bromo-2-chlorophenol 98%
4-Bromo-2-chlorophenol

Purity: 98% CAS No.: 3964-56-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Flash Point: 110°(230°F) Odor: Phenol-like Boiling Point: 232-235° Beilstein Reference: 2042867 MDL No.: MFCD00002166

Formula: C₆H₄BrClO
Boiling Pt: 232…235 °C
Melting Pt: 47…50 °C
Density: 1.617 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002166
CAS Number: 3964-56-5
EINECS: 223-572-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR