2-Chloro-α-methylbenzyl alcohol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(2-Chlorophenyl)ethanol, o-Chloro-α-methylbenzyl alcohol, 2-Chlorophenyl methyl carbinol

B24800-06 B24800-14
CAAAB24800-06EA 79.32 CAD
CAAAB24800-06 CAAAB24800-14 CAAAB24800-22
2-Chloro-α-methylbenzyl alcohol 96%
2-Chloro-α-methylbenzyl alcohol

Purity: 96% CAS No.: 13524-04-4 Molecular Formula: C8H9ClO Formula Weight: 156.61 Refractive Index: 1.5450 Boiling Point: 121-123°/14mm MDL No.: MFCD00041037

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 121…123 °C/14 mm
Storage Temperature: Ambient
MDL Number: MFCD00041037
CAS Number: 13524-04-4
EINECS: 236-868-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR