2,4,6-Trimethylthiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

B24798-03 B24798-06 B24798-14
CAAAB24798-03EA 93.54 CAD
CAAAB24798-03 CAAAB24798-06 CAAAB24798-14
2,4,6-Trimethylthiophenol 97%
2,4,6-Trimethylthiophenol

Purity: 97% CAS No.: 1541-10-2 Molecular Formula: C9H12S Formula Weight: 152.25 MDL No.: MFCD00218475

Formula: C₉H₁₂S
Storage Temperature: Ambient
MDL Number: MFCD00218475
CAS Number: 1541-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR