Benzyl butyl phthalate 98%

Supplier: Thermo Scientific Chemicals
Danger

B24769-0E B24769-22 B24769-36
CAAAAB24769-0EEA 1003.3 CAD
CAAAAB24769-0E CAAAAB24769-22 CAAAAB24769-36
Benzyl butyl phthalate 98%
Benzyl butyl phthalate

Purity: 98% CAS No.: 85-68-7 Molecular Formula: C19H20O4 Formula Weight: 312.37 Flash Point: 110°(230°F) Refractive Index: 1.5400 Boiling Point: 370° UN No.: 3082 MDL No.: MFCD00009440

Formula: C₁₉H₂₀O₄
Boiling Pt: 370 °C
Melting Pt: –35 °C
Density: 1.100
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009440
CAS Number: 85-68-7
EINECS: 201-622-7
UN: 3082
ADR: 9,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR