Methyl-2,4-dihydroxybenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24757-14
B24757-22
CAAAAB24757-06EA
37.1
CAD
CAAAAB24757-06
CAAAAB24757-14
CAAAAB24757-22
Methyl-2,4-dihydroxybenzoate 97%
Methyl 2,4-dihydroxybenzoate
Purity: 97%
CAS No.: 2150-47-2
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 116-119°
MDL No.: MFCD00002276
Formula:
C₈H₈O₄ Boiling Pt: 300 °C (1013 hPa) Melting Pt: 116…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002276 CAS Number: 2150-47-2 EINECS: 218-428-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...