2-Ethoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Danger

B24755-14 B24755-36
CAAAAB24755-14EA 33.34 CAD
CAAAAB24755-14 CAAAAB24755-22 CAAAAB24755-36
2-Ethoxyphenol 98%
2-Ethoxyphenol

Purity: 98% CAS No.: 94-71-3 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 86°(187°F) Odor: Phenol-like Boiling Point: 214-216° MDL No.: MFCD00002187

Formula: C₈H₁₀O₂
Boiling Pt: 214…216 °C
Melting Pt: 24…27 °C
Density: 1.10
Flash Pt: 86 °C(187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR