α-Bromo-m-xylene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methylbenzyl bromide

B24731-06 B24731-14
CAAAAB24731-06EA 51.35 CAD
CAAAAB24731-06 CAAAAB24731-14
α-Bromo-m-xylene 97%
α-Bromo-m-xylene

Purity: 97% CAS No.: 620-13-3 Molecular Formula: C8H9Br Formula Weight: 185.07 Storage Temperature: 2°C to 8°C Flash Point: 82°(179°F) Refractive Index: 1.5690 Boiling Point: 184-185°/340mm Beilstein Reference: 2076425 Merck Reference: 14,10090 UN No.: 1701 MDL No.: MFCD00000177

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 184…185 °C/340 mm
Melting Pt: 18 °C
Density: 1.370
Flash Pt: 82 °C (179 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000177
CAS Number: 620-13-3
EINECS: 210-625-2
UN: 1701
ADR: 6.1,II
Merck Index: 14,10090

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR