4-Amino-3,5-dichloropyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

B24725-06 B24725-14 B24725-22
CAAAB24725-06EA 83.27 CAD
CAAAB24725-06 CAAAB24725-14 CAAAB24725-22
4-Amino-3,5-dichloropyridine 98+%
4-Amino-3,5-dichloropyridine

Purity: 98+% CAS No.: 22889-78-7 Molecular Formula: C5H4Cl2N2 Formula Weight: 163.01 Melting Point: 159-163° Beilstein Reference: 120547

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00125023
CAS Number: 22889-78-7
EINECS: 245-304-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR