(±)-Nipecotic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid
B24723-03
B24723-06
B24723-14
CAAAAB24723-03EA
52.77
CAD
CAAAAB24723-03
CAAAAB24723-06
CAAAAB24723-14
(±)-Nipecotic acid 98%
(±)-Nipecotic acid
GABA reuptake inhibitor
Purity: 98%
CAS No.: 498-95-3
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: ca 261° dec.
Solubility: Soluble in water
Beilstein Reference: 81096
Merck Reference: 14,6561
MDL No.: MFCD00005992
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: ∼ 261 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005992 CAS Number: 498-95-3 EINECS: 207-873-9 Merck Index: 14,6561 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...