1,8-Octanedithiol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,8-Dimercaptooctane

B24693-03 B24693-06 B24693-14
CAAAB24693-03EA 55.62 CAD
CAAAB24693-03 CAAAB24693-06 CAAAB24693-14
1,8-Octanedithiol 98%
1,8-Octanedithiol

Purity: 98% CAS No.: 1191-62-4 Molecular Formula: HS(CH2)8SH Formula Weight: 178.36 Flash Point: 113°(235°F) Refractive Index: 1.5050 Odor: Unpleasant Boiling Point: 269-270° Beilstein Reference: 1735431 MDL No.: MFCD00003574

Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 269…270 °C
Density: 0.960
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003574
CAS Number: 1191-62-4
EINECS: 214-738-8
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR